2-(Methylsulphanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Methylthio)-1,4,5,6-tetrahydropyrimidine hydroiodide

OR23426-1G OR23426-5G
APOSOR23426-1GEA 36.4 EUR
APOSOR23426-1G APOSOR23426-5G
2-(Methylsulphanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide
2-(Methylsulphanyl)-1,4,5,6-tetrahydropyrimidinehydroiodide
Formel: C₅H₁₁IN₂S
Molecular Weight: 258,13 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00829842
CAS-Nummer: 5445-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR