4-Chlorsalicylsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-2-hydroxybenzoic acid
OR2357-25G
OR2357-100G
APOSOR2357-25GEA
21.9
EUR
APOSOR2357-25G
APOSOR2357-100G
4-Chlorsalicylsäure 98%
4-Chlorsalicylsäure
Formel:
C₇H₅ClO₃ Molecular Weight: 172,57 g/mol Schmelzpunkt: 209…213 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002449 CAS-Nummer: 5106-98-9 EINECS: 225-829-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...