2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(1,4,5,6-Tetrahydropyrimidin-2-yl)benzenethiol

OR23587-1G OR23587-250MG
APOSOR23587-1GEA 217 EUR
APOSOR23587-1G APOSOR23587-250MG
2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol
2-(1,4,5,6-Tetrahydro-2-pyrimidinyl)thiophenol
Formel: C₁₀H₁₂N₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00052612
CAS-Nummer: 53440-32-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR