Trichloroacrylic acid

Lieferant: Apollo Scientific
Gefahr

OR23621-1G OR23621-250MG
APOSOR23621-1GEA 560 EUR
APOSOR23621-1G APOSOR23621-250MG
Trichloroacrylic acid
Trichloroacrylic acid
Formel: C₃HCl₃O₂
Schmelzpunkt: 72…74 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00015455
CAS-Nummer: 2257-35-4
EINECS: 218-856-0
UN: 3261
ADR: 8,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR