4-Chlor-3-nitrobenzonitril
Lieferant: Apollo Scientific
OR23758-100G
APOSOR23758-100GEA
68.3
EUR
APOSOR23758-100G
4-Chlor-3-nitrobenzonitril
4-Chlor-3-nitrobenzonitril
Formel:
C₇H₃ClN₂O₂ Molecular Weight: 182,57 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00016987 CAS-Nummer: 939-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...