4-Amino-3,5-dimethyl-1,2,4-triazol
Lieferant: Apollo Scientific
|
Achtung
|
OR23833-1G
OR23833-5G
APOSOR23833-1GEA
72.5
EUR
APOSOR23833-1G
APOSOR23833-5G
4-Amino-3,5-dimethyl-1,2,4-triazol
4-Amino-3,5-dimethyl-1,2,4-triazole
Formel:
C₄H₈N₄ Molecular Weight: 112,14 g/mol Schmelzpunkt: 196…198 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051647 CAS-Nummer: 3530-15-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...