4-Imidazol-1-yl-phenylamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(1H-Imidazol-1-yl)aniline

OR2388-25G
APOSOR2388-25GEA 166 EUR
APOSOR2388-25G
4-Imidazol-1-yl-phenylamine
4-Imidazol-1-yl-phenylamine
Formel: C₉H₉N₃
Molecular Weight: 159.1900 g/mol
Schmelzpunkt: 143…147 °C
Lagertemperatur: Kühlschrank
MDL: MFCD01074865
CAS-Nummer: 2221-00-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR