Myosmine 98%

Lieferant: Apollo Scientific

Synonyme: 3-(3,4-dihydro-2H-pyrrol-5-yl)pyridine

OR23912-1G OR23912-5G OR23912-10G
APOSOR23912-1GEA 306 EUR
APOSOR23912-1G APOSOR23912-5G APOSOR23912-10G
Myosmine 98%
Myosmine
Formel: C₉H₁₀N₂
Molecular Weight: 146,193 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052019
CAS-Nummer: 532-12-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR