Furan-2-carbothioamide

Lieferant: Apollo Scientific
Achtung

OR23914-1G OR23914-250MG OR23914-500MG OR23914-2.5G OR23914-5G
APOSOR23914-1GEA 119 EUR
APOSOR23914-1G APOSOR23914-250MG APOSOR23914-500MG APOSOR23914-2.5G APOSOR23914-5G
Furan-2-carbothioamide
Furan-2-carbothioamide
Formel: C₅H₅NOS
Molecular Weight: 127.17 g/mol
Schmelzpunkt: 130…132 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01935200
CAS-Nummer: 17572-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR