5-(2,4-Dichlorophenyl)cyclohexane-1,3-dione

Lieferant: Apollo Scientific
Achtung

OR23983-1G OR23983-250MG OR23983-500MG OR23983-2.5G
APOSOR23983-1GEA 314 EUR
APOSOR23983-1G APOSOR23983-250MG APOSOR23983-500MG APOSOR23983-2.5G
5-(2,4-Dichlorophenyl)cyclohexane-1,3-dione
5-(2,4-Dichlorophenyl)cyclohexane-1,3-dione
Formel: C₁₂H₁₀Cl₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00104795
CAS-Nummer: 55579-70-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR