2,2'-Dipyridyldisulfid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,2'-Dithiodipyridin
OR24005-100G
OR24005-25G
OR24005-5G
APOSOR24005-100GEA
108
EUR
APOSOR24005-100G
APOSOR24005-25G
APOSOR24005-5G
2,2'-Dipyridyldisulfid
2,2'-Dipyridyldisulfid
Formel:
C₁₀H₈N₂S₂ Molecular Weight: 220,32 g/mol Schmelzpunkt: 56…58 °C Flammpunkt: 210 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00006287 CAS-Nummer: 2127-03-9 EINECS: 218-343-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...