2-Nitro-4-thiophencarbonitril

Lieferant: Apollo Scientific
Achtung

OR24042-1G OR24042-250MG
APOSOR24042-1GEA 98.6 EUR
APOSOR24042-1G APOSOR24042-250MG
2-Nitro-4-thiophencarbonitril
2-Nitro-4-thiophencarbonitril
Formel: C₅H₂N₂O₂S
Molecular Weight: 154,15 g/mol
Schmelzpunkt: 121…124 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052580
CAS-Nummer: 42137-23-5
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR