5-(2-Nitrophenyl)oxazol
Lieferant: Apollo Scientific
|
Achtung
|
OR24051-1G
OR24051-500MG
APOSOR24051-1GEA
271
EUR
APOSOR24051-1G
APOSOR24051-500MG
5-(2-Nitrophenyl)oxazol
5-(2-Nitrophenyl)oxazole
Formel:
C₉H₆N₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00085146 CAS-Nummer: 89808-75-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...