5-Oxo-1-(thien-2-ylmethyl)pyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific
Achtung

OR24057-1G OR24057-5G
APOSOR24057-1GEA 50.9 EUR
APOSOR24057-1G APOSOR24057-5G
5-Oxo-1-(thien-2-ylmethyl)pyrrolidine-3-carboxylic acid
5-Oxo-1-(thien-2-ylmethyl)pyrrolidine-3-carboxylic acid
Formel: C₁₀H₁₁NO₃S
Molecular Weight: 225,27 g/mol
Schmelzpunkt: 144…146 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00084927
CAS-Nummer: 175136-92-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR