2-(4-Chlorphenyl)thioacetamid

Lieferant: Apollo Scientific
Achtung

OR24099-1G OR24099-25G OR24099-5G
APOSOR24099-1GEA 21.9 EUR
APOSOR24099-1G APOSOR24099-25G APOSOR24099-5G
2-(4-Chlorphenyl)thioacetamid
2-(4-Chlorphenyl)thioacetamid
Formel: C₈H₈ClNS
Molecular Weight: 185,67 g/mol
Schmelzpunkt: 129…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00278249
CAS-Nummer: 17518-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR