Ethyl-2-amino-5-nitrobenzoat 98%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-amino-5-nitrobenzoate

OR24114-25G
APOSOR24114-25GEA 127 EUR
APOSOR24114-25G
Ethyl-2-amino-5-nitrobenzoat 98%
Ethyl-2-amino-5-nitrobenzoat
Formel: C₈H₈N₂O₄
Molecular Weight: 196,1601 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00278157
CAS-Nummer: 3816-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR