3,4-Dimethylbenzoesäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR24119-100G
OR24119-500G
APOSOR24119-100GEA
106
EUR
APOSOR24119-100G
APOSOR24119-500G
3,4-Dimethylbenzoesäure 98%
3,4-Dimethylbenzoic acid
Formel:
C₉H₁₀O₂ Molecular Weight: 150,18 g/mol Schmelzpunkt: 163…165 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002524 CAS-Nummer: 619-04-5 EINECS: 210-576-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...