6-Piperazin-1-ylnicotinonitrile 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-(Piperazin-1-yl)nicotinonitrile
OR2418-1G
OR2418-5G
APOSOR2418-1GEA
98.6
EUR
APOSOR2418-1G
APOSOR2418-5G
6-Piperazin-1-ylnicotinonitrile 95%
6-Piperazin-1-ylnicotinonitrile
Formel:
C₁₀H₁₂N₄ Molecular Weight: 188,23 g/mol Schmelzpunkt: 75…78 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00833024 CAS-Nummer: 149554-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...