5-Hydrazino-1,3-dimethyl-4-nitro-1H-pyrazole

Lieferant: Apollo Scientific

OR24189-1G
APOSOR24189-1GEA 184 EUR
APOSOR24189-1G
5-Hydrazino-1,3-dimethyl-4-nitro-1H-pyrazole
5-Hydrazino-1,3-dimethyl-4-nitro-1H-pyrazole
Formel: C₅H₉N₅O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00052535
CAS-Nummer: 66971-55-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR