5-Chlor-2-hydroxybenzonitril 95+%

Lieferant: Apollo Scientific
Achtung

OR24196-1G OR24196-5G
APOSOR24196-1GEA 27.7 EUR
APOSOR24196-1G APOSOR24196-5G
5-Chlor-2-hydroxybenzonitril 95+%
5-Chlor-2-hydroxybenzonitril
Formel: C₇H₄ClNO
Molecular Weight: 153.5700 g/mol
Schmelzpunkt: 152…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00219003
CAS-Nummer: 13589-72-5
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR