3-Amino-4-brom-1H-pyrazol

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Bromo-1H-pyrazol-5-amine, 4-Bromo-1H-pyrazol-3-amine, 3-Amino-4-bromopyrazole

OR24239-1G OR24239-5G OR24239-10G
APOSOR24239-1GEA 21.9 EUR
APOSOR24239-1G APOSOR24239-5G APOSOR24239-10G
3-Amino-4-brom-1H-pyrazol
3-Amino-4-bromo-1H-pyrazole
Formel: C₃H₄BrN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00082728
CAS-Nummer: 16461-94-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR