Methyl-3-amino-5-phenyl-2-thiophencarboxylat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3-amino-5-phenylthiophene-2-carboxylate

OR24261-25G
APOSOR24261-25GEA 145 EUR
APOSOR24261-25G
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Methyl-3-amino-5-phenyl-2-thiophencarboxylat
Formel: C₁₂H₁₁NO₂S
Molecular Weight: 233,2862 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00068161
CAS-Nummer: 100063-22-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR