Methyl-4-aminothiophene-3-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 4-aminothiophene-3-carboxylate
OR24350-1G
OR24350-5G
APOSOR24350-1GEA
197
EUR
APOSOR24350-1G
APOSOR24350-5G
Methyl-4-aminothiophene-3-carboxylate
Methyl-4-aminothiophene-3-carboxylate
Formel:
C₆H₇NO₂S Schmelzpunkt: 35…38 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00130102 CAS-Nummer: 69363-85-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...