3-Coumaranon

Lieferant: Apollo Scientific
Achtung

Synonyme: Benzofuran-3(2H)-one

OR24591-1G OR24591-25G OR24591-5G
APOSOR24591-1GEA 21.9 EUR
APOSOR24591-1G APOSOR24591-25G APOSOR24591-5G
3-Coumaranon
3-Coumaranon
Formel: C₈H₆O₂
Molecular Weight: 134,13 g/mol
Schmelzpunkt: 97…103 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00051810
CAS-Nummer: 7169-34-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR