(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Butyl-5-chloro-4-hydroxymethylimidazole, (2-Butyl-4-chloro-1H-imidazol-5-yl)methanol
OR2460-1G
OR2460-25G
OR2460-5G
APOSOR2460-1GEA
43.5
EUR
APOSOR2460-1G
APOSOR2460-25G
APOSOR2460-5G
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol
(2-Butyl-5-chloro-1H-imidazol-4-yl)methanol
Formel:
C₈H₁₃ClN₂O Molecular Weight: 188.66 g/mol Schmelzpunkt: 150…152 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01934397 CAS-Nummer: 79047-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...