1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-(Aminomethyl)-2,3-dihydrobenzo[b]furan
OR2467-1G
OR2467-5G
APOSOR2467-1GEA
58
EUR
APOSOR2467-1G
APOSOR2467-5G
1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine
1-(2,3-Dihydro-1-benzofuran-5-yl)methanamine
Formel:
C₉H₁₁NO Molecular Weight: 149.19 g/mol Siedepunkt: 98…100 °C/0.1mmHg Schmelzpunkt: 38…40 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00728869 CAS-Nummer: 55745-74-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...