5-Amino-4,6-dichlor-2-methylpyrimidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-dichloro-2-methylpyrimidin-5-amine

OR24758-25G OR24758-5G
APOSOR24758-25GEA 33.5 EUR
APOSOR24758-25G APOSOR24758-5G
5-Amino-4,6-dichlor-2-methylpyrimidin
5-Amino-4,6-dichloro-2-methylpyrimidine
Formel: C₅H₅Cl₂N₃
Molecular Weight: 178,0200 g/mol
Schmelzpunkt: 70…75 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00194053
CAS-Nummer: 39906-04-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR