2-Butylimidazol-4-carbaldehyd

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Butyl-1H-imidazole-5-carboxaldehyde

OR2486-1G OR2486-250MG
APOSOR2486-1GEA 27.7 EUR
APOSOR2486-1G APOSOR2486-250MG
2-Butylimidazol-4-carbaldehyd
2-Butylimidazol-4-carbaldehyd
Formel: C₈H₁₂N₂O
Molecular Weight: 152,193 g/mol
Schmelzpunkt: 117…120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01075030
CAS-Nummer: 68282-49-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR