4-Bromo-1H-pyrazole-5-carbonitrile
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Bromo-1H-pyrazole-3-carbonitrile
OR2487-1G
OR2487-250MG
OR2487-5G
APOSOR2487-1GEA
87
EUR
APOSOR2487-1G
APOSOR2487-250MG
APOSOR2487-5G
4-Bromo-1H-pyrazole-5-carbonitrile
4-Bromo-1H-pyrazole-5-carbonitrile
Formel:
C₄H₂BrN₃ Molecular Weight: 171.98 g/mol Schmelzpunkt: 154…160 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00105683 CAS-Nummer: 288246-16-2 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...