2-Methyl-4-nitroindole

Lieferant: Apollo Scientific
Achtung

OR2507-100MG OR2507-500MG
APOSOR2507-100MGEA 21.9 EUR
APOSOR2507-100MG APOSOR2507-500MG
2-Methyl-4-nitroindole
2-Methyl-4-nitroindole
Formel: C₉H₈N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD09028445
CAS-Nummer: 3484-10-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR