Ethyl 3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo-[c]thiophene-1-carboxylate

Lieferant: Apollo Scientific

OR25278-10G OR25278-1G OR25278-5G
APOSOR25278-10GEA 260 EUR
APOSOR25278-10G APOSOR25278-1G APOSOR25278-5G
Ethyl 3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo-[c]thiophene-1-carboxylate
Ethyl-3-(butylthio)-4-oxo-4,5,6,7-tetrahydrobenzo[c]thiophene-1-carboxylate
Formel: C₁₅H₂₀O₃S₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00085061
CAS-Nummer: 172516-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR