5-Chlor-1-methyl-4-nitro-1H-imidazol

Lieferant: Apollo Scientific
Achtung

OR25467-100G OR25467-25G OR25467-500G
APOSOR25467-100GEA 65.4 EUR
APOSOR25467-100G APOSOR25467-25G APOSOR25467-500G
5-Chlor-1-methyl-4-nitro-1H-imidazol
5-Chloro-1-methyl-4-nitroimidazole
Formel: C₄H₄ClN₃O₂
Molecular Weight: 161,55 g/mol
Schmelzpunkt: 148…150 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00233664
CAS-Nummer: 4897-25-0
EINECS: 225-521-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR