Methyl-2,4-dichloropyrimidine-6-carboxylate
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichloro-4-pyrimidinecarboxylic acid methyl ester, 2-(3,4-dichlorophenyl)-4-methyl-5-thiazolecarboxylic acid, Methyl 2,6-dichloro-4-pyrimidinecarboxylate, Methyl 2,6-dichloropyrimidine-4-carboxylate
OR2558-25G
OR2558-1G
OR2558-5G
APOSOR2558-25GEA
109
EUR
APOSOR2558-25G
APOSOR2558-1G
APOSOR2558-5G
Methyl-2,4-dichloropyrimidine-6-carboxylate
Methyl-2,4-dichloropyrimidine-6-carboxylate
Formel:
C₆H₄Cl₂N₂O₂ Molecular Weight: 207,02 g/mol Schmelzpunkt: 54…56 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941317 CAS-Nummer: 6299-85-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...