4-Phenylethynylbenzaldehyde 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-(Phenylethynyl)benzaldehyde

OR25608-1G OR25608-5G OR25608-25G
APOSOR25608-1GEA 33.5 EUR
APOSOR25608-1G APOSOR25608-5G APOSOR25608-25G
4-Phenylethynylbenzaldehyde 95%
4-Phenylethynylbenzaldehyde
Formel: C₁₅H₁₀O
Molecular Weight: 206,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00114585
CAS-Nummer: 57341-98-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR