1-Acetyl-5-bromindolin

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-(5-Bromoindolin-1-yl)ethanone

OR25617-1G OR25617-25G
APOSOR25617-1GEA 78.3 EUR
APOSOR25617-1G APOSOR25617-25G
1-Acetyl-5-bromindolin
1-Acetyl-5-bromindolin
Formel: C₁₀H₁₀BrNO
Molecular Weight: 240,1 g/mol
Schmelzpunkt: 118…121 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00056017
CAS-Nummer: 22190-38-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR