4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol 95+%

Lieferant: Apollo Scientific

OR2563-10G OR2563-1G OR2563-5G
APOSOR2563-10GEA 667 EUR
APOSOR2563-10G APOSOR2563-1G APOSOR2563-5G
4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol 95+%
4-[(4,6-Dimethylpyrimidin-2-yl)thio]phenol
Formel: C₁₂H₁₂N₂OS
Schmelzpunkt: 192…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00203057
CAS-Nummer: 107718-34-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR