6,7-Dimethoxy-4-chinazolon
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6,7-Dimethoxy-4(3H)-quinazolinone, 6,7-Dimethoxy-1H-quinazolin-4-one
OR2564-5G
OR2564-25G
OR2564-100G
APOSOR2564-5GEA
55.1
EUR
APOSOR2564-5G
APOSOR2564-25G
APOSOR2564-100G
6,7-Dimethoxy-4-chinazolon
6,7-Dimethoxy-4-chinazolon
Formel:
C₁₀H₁₀N₂O₃ Molecular Weight: 206.2 g/mol Schmelzpunkt: 298 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01570147 CAS-Nummer: 13794-72-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...