3-[(2-Furylmethyl)amino]propanenitrile

Lieferant: Apollo Scientific

OR25697-1G OR25697-5G
APOSOR25697-1GEA 0 EUR
APOSOR25697-1G APOSOR25697-5G
3-[(2-Furylmethyl)amino]propanenitrile
3-[(2-Furylmethyl)amino]propanenitrile
Formel: C₈H₁₀N₂O
Molecular Weight: 150,18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00219840
CAS-Nummer: 6788-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR