2-Brom-4-nitroimidazol 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Brom--5-nitro1H-imidazol

OR2576-10G OR2576-5G
APOSOR2576-10GEA 40.6 EUR
APOSOR2576-10G APOSOR2576-5G
2-Brom-4-nitroimidazol 97%
2-Bromo-4-nitroimidazole
Formel: C₃H₂BrN₃O₂
Molecular Weight: 191,97 g/mol
Schmelzpunkt: 235 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09025308
CAS-Nummer: 65902-59-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR