2-Chloro-4-nitro-1H-imidazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-5-nitro-1H-imidazole
OR2577-1G
OR2577-25G
OR2577-5G
APOSOR2577-1GEA
21.9
EUR
APOSOR2577-1G
APOSOR2577-25G
APOSOR2577-5G
2-Chloro-4-nitro-1H-imidazole
2-Chloro-4-nitro-1H-imidazole
Formel:
C₃H₂ClN₃O₂ Molecular Weight: 147.518 g/mol Schmelzpunkt: 214…216 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03419295 CAS-Nummer: 57531-37-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...