3-[4-(Acetylamino)phenyl]acrylic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: trans-4-(Acetylamino)cinnamic acid

OR25793-1G OR25793-5G
APOSOR25793-1GEA 58 EUR
APOSOR25793-1G APOSOR25793-5G
3-[4-(Acetylamino)phenyl]acrylic acid
3-[4-(Acetylamino)phenyl]acrylic acid
Formel: C₁₁H₁₁NO₃
Molecular Weight: 205,22 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00016846
CAS-Nummer: 7152-04-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR