5-Chlor-3-pyridinboronsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Chloropyridine-3-boronic acid, (5-Chloropyridin-3-yl)boronic acid
OR2582-1G
OR2582-25G
OR2582-5G
APOSOR2582-1GEA
23.2
EUR
APOSOR2582-1G
APOSOR2582-25G
APOSOR2582-5G
5-Chlor-3-pyridinboronsäure
5-Chloro-3-pyridineboronic acid
Formel:
C₅H₅BClNO₂ Molecular Weight: 157,36 g/mol Schmelzpunkt: 255…260 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD06798243 CAS-Nummer: 872041-85-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...