5-Brom-2-chlor-3-pyridinboronsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Brom-2-chlorpyridin-3-boronsäure, 5-Brom-2-chlor-3-pyridinylboronsäure
OR2583-1G
OR2583-250MG
OR2583-5G
APOSOR2583-1GEA
206
EUR
APOSOR2583-1G
APOSOR2583-250MG
APOSOR2583-5G
5-Brom-2-chlor-3-pyridinboronsäure 97%
5-Bromo-2-chloro-3-pyridineboronic acid
Formel:
C₅H₄BBrClNO₂ Molecular Weight: 236.26 g/mol Schmelzpunkt: 148…150 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09037486 CAS-Nummer: 1072944-19-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...