Tetrahydroquinoline-6-carboxylic acid 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1,2,3,4-Tetrahydroquinoline-6-carboxylic acid
OR25899-1G
OR25899-5G
APOSOR25899-1GEA
154
EUR
APOSOR25899-1G
APOSOR25899-5G
Tetrahydroquinoline-6-carboxylic acid 95+%
Tetrahydroquinoline-6-carboxylic acid
Formel:
C₁₀H₁₁NO₂ Molecular Weight: 177,2 g/mol Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00957087 CAS-Nummer: 5382-49-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...