5-Acetyl-2-chlorpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Acetyl-6-chloropyridine

OR25927-1G OR25927-5G
APOSOR25927-1GEA 21.9 EUR
APOSOR25927-1G APOSOR25927-5G
5-Acetyl-2-chlorpyridin
5-Acetyl-2-chlorpyridin
Formel: C₇H₆ClNO
Molecular Weight: 155,5816 g/mol
Schmelzpunkt: 93…95 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03407343
CAS-Nummer: 55676-22-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR