(1-Benzyl-2-mercapto-1H-imidazol-5-yl)methanol

Lieferant: Apollo Scientific

OR25935-10G OR25935-1G OR25935-5G
APOSOR25935-10GEA 519 EUR
APOSOR25935-10G APOSOR25935-1G APOSOR25935-5G
(1-Benzyl-2-mercapto-1H-imidazol-5-yl)methanol
(1-Benzyl-2-mercapto-1H-imidazol-5-yl)methanol
Formel: C₁₁H₁₂N₂OS
Molecular Weight: 220,3 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02179523
CAS-Nummer: 98412-23-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR