Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat

Lieferant: Apollo Scientific

Synonyme: Ethyl 6-chloroimidazo[1,2-b]pyridazine-2-carboxylate

OR25936-5G
APOSOR25936-5GEA 79.9 EUR
APOSOR25936-5G
Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat
Ethyl-6-chlorimidazo[1,2-b]pyridazin-2-carboxylat
Formel: C₉H₈ClN₃O₂
Molecular Weight: 225.6317 g/mol
Schmelzpunkt: 155…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03086230
CAS-Nummer: 64067-99-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR