4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid

Lieferant: Apollo Scientific

Synonyme: 4-(2,2,2-Trimethylacetamino)pyridine-3-carboxylic acid

OR25946-1G OR25946-250MG OR25946-500MG OR25946-2.5G OR25946-5G
APOSOR25946-1GEA 275 EUR
APOSOR25946-1G APOSOR25946-250MG APOSOR25946-500MG APOSOR25946-2.5G APOSOR25946-5G
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid
4-[(2,2-Dimethylpropanoyl)amino]nicotinic acid
Formel: C₁₁H₁₄N₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03407344
CAS-Nummer: 125867-31-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR