3-Methoxythiophene-2-carboxylic acid 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR2595-1G
OR2595-250MG
APOSOR2595-1GEA
223
EUR
APOSOR2595-1G
APOSOR2595-250MG
3-Methoxythiophene-2-carboxylic acid 95%
3-Methoxythiophene-2-carboxylic acid
Formel:
C₆H₆O₃S Schmelzpunkt: 181…183 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01316613 CAS-Nummer: 60166-83-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...