3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbonsäure 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR25961-1G
OR25961-250MG
OR25961-5G
APOSOR25961-1GEA
174
EUR
APOSOR25961-1G
APOSOR25961-250MG
APOSOR25961-5G
3-(4-Methoxyphenyl)-5-methyl-4-isoxazolcarbonsäure 97%
3-(4-Methoxyphenyl)-5-methylisoxazole-4-carboxylic acid
Formel:
C₁₂H₁₁NO₄ Schmelzpunkt: 190…195 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03407357 CAS-Nummer: 93041-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...